Painter's Bib Overalls, Dickies®
Fournisseur: Dickies
Certificats
A propos de cet article
Dickies® style 8953.
- Roomy bib pocket with pencil slot
- Hammer loops and multiple tool pockets on both legs
- Sturdy 100% cotton painter's drill fabric is built to last
- Triple-stitched seams for strength
- 8.75 oz. painters drill, 100% cotton
Dickies® signature brass buttons at sides and suspenders. Elastic shoulder straps adjust for a perfect fit and easy on-off.
Spécifications
- Catalog No:
- 76149-078
- 76149-080
- 76149-052
- 76149-082
- 76149-084
- 76149-074
- 76149-076
- 76149-086
- 76149-088
- 76149-090
- 76149-092
- 76149-060
- 76149-058
- 76149-056
- 76149-054
- 76149-068
- 76149-066
- 76149-064
- 76149-062
- 76149-100
- 76149-072
- 76149-070
- 76149-094
- 76149-096
- 76149-098
- Model:
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- 8953
- Garment application:
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Industrial, Paint and lacquer, Services
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Fabric:
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton