Women's Comfort Waist EMT Pants, Dickies®
Fournisseur: Dickies
Certificats
A propos de cet article
Dickies® style FP2377, relaxed fit, straight leg.
- StayDark® technology
- Triple grip waistband
- Double hook & eye closure
- Reinforced double knee
- Full size functional cargo pockets
- Tool pocket with heavy-duty snap loops
- Heavy-duty brass ratcheting zipper
Made of 7.25 oz. flex twill fabric , 65% polyester/35% cotton.
Sizing Chart: UU size designations represent a 37" inseam, unhemmed.
Spécifications
- Catalog No:
- 76145-558
- 76145-602
- 76145-600
- 76145-622
- 76145-620
- 76145-624
- 76145-610
- 76145-608
- 76145-606
- 76145-628
- 76145-604
- 76145-626
- 76145-618
- 76145-616
- 76145-614
- 76145-612
- Model:
- FP2377
- FP2377
- FP2377
- FP2377
- FP2377
- FP2377
- FP2377
- FP2377
- FP2377
- FP2377
- FP2377
- FP2377
- FP2377
- FP2377
- FP2377
- FP2377
- Garment application:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Gender:
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Fabric:
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- Weight:
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.
- 7.25 oz.