Indigo Bib Overalls, Dickies®
Fournisseur: Dickies
Certificats
A propos de cet article
Dickies® style 83294.
- Zipper fly
- Cross-over high-back style
- Large bib pocket with pencil division and watch pocket
- Hammer loop and double tool pocket
- Triple stitched seams
- 11.75 oz. fine weave denim, 100% cotton
Spécifications
- Catalog No:
- 76166-232
- 76166-230
- 76166-236
- 76166-192
- 76166-234
- 76166-190
- 76166-240
- 76166-196
- 76166-238
- 76166-194
- 76166-244
- 76166-200
- 76166-242
- 76166-198
- 76166-248
- 76166-204
- 76166-246
- 76166-202
- 76166-208
- 76166-206
- 76166-250
- 76166-254
- 76166-210
- 76166-252
- 76166-214
- 76166-258
- 76166-256
- 76166-212
- 76166-218
- 76166-260
- 76166-216
- 76166-222
- 76166-220
- 76167-318
- 76166-226
- 76166-182
- 76166-224
- 76166-180
- 76167-322
- 76166-186
- 76167-320
- 76166-228
- 76166-184
- 76166-188
- Model:
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- 83294
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Chest long
- Fabric:
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Fine weave denim, 100% cotton
- Weight:
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.
- 11.75 oz.