Women's Stretch Oxford Shirts, Dickies®
Fournisseur: Dickies
Certificats
A propos de cet article
Dickies® style FL254, long sleeve.
- Mechanical stretch fabric for ease of movement
- Tailored fit
- Easy care stain release, moisture wicking, resists wrinkles
- Pearlized buttons
- Two-piece lined collar with collar stays
- Back tuck pleats, left chest pocket
- Tailored sleeve placket and cuff
Made of 5 oz. Oxford flex fabric, 60% cotton/40% polyester.
Spécifications
- Catalog No:
- 76145-120
- 76145-118
- 76145-116
- 76145-114
- 76145-122
- 76144-846
- 76144-844
- 76145-052
- 76145-050
- 76145-048
- 76145-126
- 76145-124
- 76145-054
- 76144-980
- 76145-134
- 76145-132
- 76144-978
- 76145-130
- 76144-976
- 76145-128
- 76144-982
- 76144-832
- 76144-840
- 76144-838
- 76144-836
- 76144-834
- 76145-112
- 76145-110
- 76145-108
- 76144-842
- Model:
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- FL254
- Garment application:
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Casual
- Gender:
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- 60% cotton/40% polyester
- Weight:
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.
- 5 oz.