GENIE Temp-Shaker 100™, Scientific Industries
# de catalogue 76226-090
Fournisseur: Scientific Industries
Spécifications
- Speed range:20 – 100 rpm
- Overall dimensions:450×480 mm
- Chamber volume:40 L
- Orbit:19 mm
- Chamber dimensions:305 mm High
- Platform size:305×305 mm
- Weight:9 kg
- Cat. no.:76226-090
- Electrical:120 V (US)
- Platform Capacity:3 kg
Spécifications
A propos de cet article
Gentle Orbital Shaking from 20 to 100 rpm, while providing precise temperature control from 28 to 75 °C, for use in many laboratory applications.
- Unique 3-way access clear dome chamber provides 360° view at all times
- Quick access to your samples with easy tilt-n-rest feature using the exclusive "kick stand" feature to open the chamber cover, left, right, or back, to optimize bench space
- The lightweight cover can also be easily removed for cleaning and attaching accessories
- Unique airflow design can heat the chamber to a stable 75 °C in as little as 20 minutes
- With the chamber door open, unit will maintain the chamber temperature within ±3 °C
Various voltages and plugs available for operation around the world. Product is produced and shipped from within the US.
BodyTextAccessories: No additional accessories necessary for operation of this model. However, stacking tray is available to double capacity. No tools necessary to mount accessories.
Delivery: This model includes the non-slip tray with drip-lip to contain any spills. Non-slip stacking tray and additional interchangeable platform styles available, sold as accessories.
Customers Who Bought This Also Bought