Women's Premium Cargo Pants, Dickies®
Fournisseur: Dickies
Certificats
A propos de cet article
Dickies® style FP2372, relaxed fit, straight leg.
- Resists wrinkles
- Easy care stain release
- Premium construction through rise for better shape and comfort
- Contour waistband to prevent gapping
- Two back welt pockets
- Metal tack button waist closure
- Plus size has extra darts for better fit , E-Z Touch fabric
This pant matches Men's style. It is made of 6.75 oz. flex twill fabric, 65% polyester/35% cotton.
Sizing chart: RG (Regular) size designations represent a 31.5" inseam. UU size designations represent a 37" inseam, unhemmed.
Spécifications
- Catalog No:
- 76145-162
- 76145-556
- 76145-160
- 76145-554
- 76145-420
- 76145-486
- 76145-484
- 76145-418
- 76145-416
- 76145-414
- 76145-538
- 76145-536
- 76145-534
- 76145-532
- 76145-542
- 76145-540
- 76145-470
- 76145-468
- 76145-478
- 76145-544
- 76145-476
- 76145-474
- 76145-472
- 76145-552
- 76145-550
- 76145-482
- 76145-548
- 76145-546
- 76145-480
- 76145-516
- 76145-514
- 76145-512
- 76145-510
- 76145-522
- 76145-520
- 76145-518
- 76145-448
- 76145-446
- 76145-444
- 76145-456
- 76145-454
- 76145-452
- 76145-450
- 76145-530
- 76145-528
- 76145-462
- 76145-460
- 76145-526
- 76145-458
- 76145-524
- 76145-494
- 76145-492
- 76145-490
- 76145-488
- 76145-422
- 76145-502
- 76145-500
- 76145-498
- 76145-496
- 76145-426
- 76145-424
- 76145-434
- 76145-432
- 76145-430
- 76145-428
- 76145-508
- 76145-442
- 76145-506
- 76145-440
- 76145-504
- 76145-438
- 76145-436
- Model:
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- FP2372
- Garment application:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Gender:
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- 65% polyester/35% cotton
- Weight:
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.
- 6.75 oz.