Vikan Ergonomic Shovel, Small Blade, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
The slim, long and small blade combination makes it suitable for shoveling smaller volumes and reaching into deep containers.
- Reinforced ergonomic and easy grip handle
- Wide handle with opening is ideal both for worker comfort and as a storage method
- Autoclavable at 250 °F/121 °C
- Made with FDA Compliant materials
This robust ergonomic shovel is designed for use in direct contact with food. The handle is reinforced with an inner aluminum tube. It is stepped in two places which helps to prevent back injuries. The neck joint has been strengthened in order to withstand heavy duty use.
Delivery: Dropshipping available.
Customers Who Bought This Also Bought