Roylco Sensory Paper
# de catalogue 470234-716
Fournisseur: Roylco
Spécifications
- Description:Sensory Paper
- Height:17.75"
- Cat. no.:470234-716
- Depth:0.02"
- Width:15"
- Includes:36 Sheets, Guide
- Weight:0.7 lbs.
Spécifications
A propos de cet article
White Embossed Paper in 6 Designs.
- Features 36 sheets of white embossed paper
- 6 designs
- Ideal for ages 2+
- Made in the USA
Kids love to explore with their hands. It’s an important part of the way they learn about the world. With this unique, white embossed paper, kids can feel the bumps and waves beneath their fingers as they create pictures with paint, pastels or charcoal. Circles, squares and other shapes guide little fingers over the tactile surfaces, enhancing expression and awareness. Follow the patterns to make colorful creations come to life, or connect the dots to make fun patterns of your own. There are 6 different designs to explore, each one offering crisp textures to stimulate the sense of touch.
Ordering information: Includes 36 sheets, 12×12" (30 cm), 6 designs, white embossed paper.
Packaging: Poly bag with header.
Customers Who Bought This Also Bought