Bottles, Pre-Cleaned, Square, HDPE, Environmental Express®
Supplier: Environmental Express
Certificates
About this item
Store more samples side-by-side.
- Square shape lets users take advantage of precious shelf space
- Keep a better grip and minimize dropping
- Leakproof lid keeps samples inside
Level 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory.
Level 3 containers meet EPA guidelines as well, but do not ship with a Certificate; ideal for general-use.
Customers Who Bought This Also Bought