Fine Ovens, Yamato
Fournisseur: Yamato Scientific
Certificats
A propos de cet article
Highly reliable and accurate fine ovens with simple settings for various temperature characteristic tests requiring complicated programming.
- Improved visibility and operability of control panel
- Precise temperature stability and uniformity
- Standard equipped with adjustable air speed function and displays of power consumption, CO2 emission and heat timer
- Improved safety with fan motor error detection
- Equipped with exhaust and cable ports
Safety features such as self-diagnostic functions, a key lock function, a control box switch, overheating prevention, and an electric leakage breaker with over-current protection are provided.
The interior is made of stainless steel, and an exhaust damper allows quick exhaust and cooling of the inside chamber.
The digital display illustrates the temperature in orange LED lights. The oven also features a timer, an integration-time function, and calibration offset.
Spécifications
- Catalog No:
- 76209-650
- 76209-648
- 76209-646
- Control type:
- Digital PID Controller
- Digital PID Controller
- Digital PID Controller
- Exhaust ports:
- 1 piece (Rear)
- 1 piece (Rear)
- 1 piece (Rear)
- No. of shelf rest steps/pitch:
- 9 Steps/45 mm
- 9 Steps/60 mm
- 9 Steps/45 mm