Programmable Refrigerant Incubators, Yamato
Supplier: Yamato Scientific
Certificates
About this item
Standard low temperature forced convection incubators applicable for low temperature tests and environmental tests.
- Inner glass door keeps temperature stable during sample observation
- Designed with a large dual glass door and inner door that forms a triplex glass door for improved heat retention (Models IN604W/804W)
- Interior light for better sample visibility (Models IN604W/804W)
- Optional slide shaker table available to put in and take out samples easily (Models IN604W/804W)
- Digital PID Controller
It has an operating temperature range of –10 to+50 °C and has high accuracy temperature control and temperature distribution.
Specifications
- Catalog No:
- 76209-634
- 76209-632
- 76209-630
- 76209-628
- 76209-626
- 76209-624
- 76209-622
- 76209-620
- 76209-618
- 76209-636
- Temperature setting accuracy:
- ±0.3 °C (refrigerator in continuous operation)
±1.0 °C (refrigerator in cycle operation) - ±0.3 °C (refrigerator in continuous operation)
±1.0 °C (refrigerator in cycle operation) - ±0.3 °C (refrigerator in continuous operation)
±1.0 °C (refrigerator in cycle operation) - ±0.3 °C (refrigerator in continuous operation)
±1.0 °C (refrigerator in cycle operation) - ±0.3 °C (refrigerator in continuous operation)
±1.0 °C (refrigerator in cycle operation) - ±0.3 °C (refrigerator in continuous operation)
±1.0 °C (refrigerator in cycle operation) - ±0.3 °C (refrigerator in continuous operation)
±1.0 °C (refrigerator in cycle operation) - ±0.3 °C (refrigerator in continuous operation)
±1.0 °C (refrigerator in cycle operation) - ±0.3 °C (refrigerator in continuous operation)
±1.0 °C (refrigerator in cycle operation) - ±0.3 °C (refrigerator in continuous operation)
±1.0 °C (refrigerator in cycle operation)
- Temperature range:
- −10…50 °C
- −10…50 °C
- −10…50 °C
- −10…50 °C
- −10…50 °C
- −10…50 °C
- −10…50 °C
- −10…50 °C
- −10…50 °C
- −10…50 °C
- No. of shelf rest steps/pitch:
- 13 Steps/30 mm
- 23 Steps/30 mm
- 23 Steps/30 mm
- 13 Steps/30 mm
- 13 Steps/30 mm
- 23 Steps/30 mm
- 23 Steps/30 mm
- 13 Steps/30 mm
- 13 Steps/30 mm
- 13 Steps/30 mm