Medline® Bath Towels
# de catalogue 470303-250
Fournisseur: MEDLINE INDUSTRIES, INC.
Spécifications
- Length:48"
- Width:24"
- Color:White
- Description:Bath Towels
- Cat. no.:470303-250
Spécifications
A propos de cet article
Soft, durable and affordable.
- Rapidly drying
- Made of 85/15 cotton polyester blend
- Treated with hydrogen peroxide for a brighter white appearance
Help patients feel more comfortable with these soft and absorbent bath towels. The fibers are woven to provide a long service life and treated with peroxide to provide a bright white color.
Customers Who Bought This Also Bought