Medline® Pocket Otoscope
# de catalogue 470303-258
Fournisseur: MEDLINE INDUSTRIES, INC.
Spécifications
- Description:Pocket otoscope, black
- Cat. no.:470303-258
Spécifications
A propos de cet article
Small and powerful.
- Compact AA size
- Consistent light output
- Brilliant LED light source
This pocket otoscope has all the features you would expect on larger models, with the convenience of an AA sized handle. Unlike some pocket-sized scopes, this otoscope has a metal housing and a glass viewing window. The 3.7 V LED provides consistent illumination throughout the 50-hour battery life. The small size fits perfectly in a pocket, or a field bag. If you want some extra travel protection it even comes with a hard-shell case to prevent damage from harder shocks.
Ordering information: An insufflation bulb is not included. If you would like to purchase one, please contact customer service. Does not include batteries.
Customers Who Bought This Also Bought