Medline® Otoscope/Ophthalmoscope Desk Set
# de catalogue 470303-256
Fournisseur: MEDLINE INDUSTRIES, INC.
Spécifications
- Description:Diagnostic Desk Set
- Cat. no.:470303-256
Spécifications
A propos de cet article
Designed for demanding everyday use.
- LED illumination for otoscopes
- 3.5 V xenon illumination for ophthalmological instruments supply daylight conditions
- With desktop charger
Always have your instruments handy with this convenient desktop Otoscope and Ophthalmoscope bundle. The slim AA sized handles are easy to grip and fit right in your pocket out of the way. When they aren't being used they store easily in the compact charging station, so they are always ready.
The ophthalmoscope uses a diopter wheel with 29 corrective lenses, an easy to operate aperture wheel and a xenon bulb for the best possible illumination. The Otoscope features a 3X magnification swiveling optical glass, and a Fiberoptic LED light source.
Customers Who Bought This Also Bought