Medline® Wall Mounted Diagnostic Set
# de catalogue 470303-254
Fournisseur: MEDLINE INDUSTRIES, INC.
Spécifications
- Description:Wall Mount Diagnostic Set
- Cat. no.:470303-254
Spécifications
A propos de cet article
A necessity for exam training.
- LED illumination for otoscope, and xenon for ophthalmoscope
- Wall mounted convenience
- Long three-meter spiral instrument cord
The wall mounted diagnostic set can be conveniently placed in exam rooms and expanded to the left or right as desired. This kit includes a power supply, a LED Otoscope and a Xenon Ophthalmoscope. Instruments automatically switch on when removed from the cradle, and off when replaced. A consistent power source extends bulb life and removes the need for constant battery changes.
Customers Who Bought This Also Bought