Small Capacity Glas-Col® Stirmantle, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Mantle stirring system with coil induced Synchostart magnetic field, exclusive to Glas-Col.
- Variable speed StirControl II controller has a 4 foot cord and can be placed outside the hood
- StirControl II features two receptacles for operating two Stirmantles at once
- Complete system includes mantle, StirControl II, PTFE stirbar, and cord
Stirmantle with cords only does not include StirControl II or PTFE stirbar, for heating needs.
Ordering information: 230 V products are not returnable.
Customers Who Bought This Also Bought