Dangerous Goods UN EPS Foam Packaging, Qorpak
# de catalogue 16465-066
Fournisseur: Qorpak
Small Business Enterprise
Spécifications
- Capacity:4 L
- Description:UN EPS foam packaging with 4 L amber glass jug and phenolic cap
- Cat. no.:16465-066
- UN Markings:4G/X 9.1/S
- Supplier No.:HMS-6551M
Spécifications
A propos de cet article
EPS Foam provides better protection and cushioning than standard fibreboard packaging.
Certifications: Meets UN 4G standards for Packing Groups II and III.
Ordering information: This package contains a pressure tested amber plastic coated glass jug and phenolic cap. For air shipments, check the regulations for any absorbent requirements and quantity limitations. Includes LDPE bag. Absorbent is sold separately.
Packaging: Sold in a convenience pack of 4.
Customers Who Bought This Also Bought