OmegaZen™ Pipette Controller, Argos Technologies®
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Precisely and rapidly pipette liquids with a comfortable, ergonomic design.
- Easily fine-tune pipetting speed with thumbwheel control and speedometer display
- Quick and quiet pump fills a 25 ml pipette in 3 seconds
- Accepts glass or plastic serological pipettes, from 1 to 100 ml
- Durable, UV-resistant plastic housing is ideal for use in cleanrooms and fume hoods
- Avoid battery depletion with auto shut-off after 5 minutes of inactivity
- Removable nosepiece and silicone adapter are autoclavable at 121 °C
BodyTextAccessories: Four accessory filter options are available to choose from. All have a polypropylene housing and hydrophobic PTFE membrane. Filter should be replaced every three months, when the filter is contaminated, or suction capacity is slowed.
Ordering information: Each Pipette Controller includes two 0.45 µm sterile filters, nosepiece, silicone adapter, wall-mount holder, rechargeable lithium-ion battery, charging desk stand, and AC adapter.
Spécifications
- Catalog No:
- 76218-692
- 76218-688
- Compatible with:
- 1 to 100 mL Glass or Plastic Serological Pipettes
- 1 to 100 mL Glass or Plastic Serological Pipettes
- Dispensing speed:
- 9 to 10 mL/s
- 9 to 10 mL/s
- No. of speeds:
- 8
- 8
- Power:
- 100 to 240 VAC
- 100 to 240 VAC
- Filter type:
- 0.45 µm Sterile PTFE
- 0.45 µm Sterile PTFE
- Body material:
- UV-resistant plastic
- UV-resistant plastic
- Battery type:
- 3.7 V Lithium Ion, Rechargeable
- 3.7 V Lithium Ion, Rechargeable