Lab Stand Support Shelf, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Lab stand support shelf fits any stand with up to 5/8" O.D. rod. Rod-mounted, and features a 5/8" O.D. × 12" long stainless rod with a through hole.
- Manufactured from light, cast aluminum and epoxy coated
Small platform is for use with up to #50 Ace-Thred adapters. Large platform is for use with up to #80 Ace-Thred adapters.
Customers Who Bought This Also Bought