Densito and DensitoPro Handheld Density Meters, METTLER TOLEDO®
Fournisseur: Mettler Toledo
Certificats
A propos de cet article
Designed for use in the lab and on the go, the Densito and DensitoPro are perfect for all applications.
- Perfect for quality control of incoming goods to at-line quality assurance of intermediate or final products
- Users can be guided through multi-step procedures with clear directions, from measurement to cleaning
BodyTextAccessories: A wide assortment of accessories are available, but must be ordered separately. These include a standard filling tube, as well as a connector syringe/filling tube for use with viscous samples. A carrying case, USB-C charging cable, and power supply with worldwide adapters are also available.
Spécifications
- Catalog No:
- 76213-948
- 76213-950
- Measurement range (°Brix):
- 0.000 – 3.000 g/cm3
- 0.000 – 3.000 g/cm3
- Measurement range:
- 0.000 – 3.000 g/cm3
- 0.000 – 3.000 g/cm3
- Temperature accuracy:
- ±0.2 °C
- ±0.2 °C
- Temperature range:
- 0…50 °C (–10…+50 °C ambient range)
- 0…50 °C (–10…+50 °C ambient range)
- Temperature resolution:
- 0.1 °C
- 0.1 °C
- Sample size:
- 2 mL
- 2 mL