Vikan® Foam Blade Squeegee, 24" Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
Excellent on all types of floors, including the grouting between tiles.
- Made with FDA compliant materials
- Autoclavable at 121 °C (250 °F)
The foam rubber blades on this squeegee make it the most effective model in the Vikan® range. It is excellent on all types of floors, as the soft rubber reaches into and cleans all spaces on the floor, such as grouting between tiles. The cartridge system blades can be removed for easy cleaning and replaced when worn out, making the model a very economical option, The foam blade squeegee may be used with any European-thread handle.
BodyTextAccessories: Replacement squeegee blades available, see 76008-210 or 76008-212.
Delivery: Dropshipping available.
Customers Who Bought This Also Bought