Vikan® Double Blade Ultra Hygiene Squeegee, 20" Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
The double blade, non-porous rubber design of this squeegee is effective in removing water.
- Made with FDA compliant materials
- Autoclavable at 121 °C (250 °F)
The double blade, non-porous rubber design of this squeegee is effective in removing water from all types of floors; tiles, epoxy, or cement. The cartridge system blades can be removed for easy cleaning and replaced when worn out, making the model a very economical hygienic option.
BodyTextAccessories: Replacement squeegee blades available, see 76006-692, 76006-694, 76006-696, 76006-698, 76008-388.
Delivery: Dropshipping available.
Customers Who Bought This Also Bought