Vikan® Single Blade Ultra Hygiene Squeegee, 24", Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
One-piece construction is effective in areas requiring the highest level of hygiene.
- Made with FDA compliant materials
- Autoclavable at 121 °C (250 °F)
The Vikan® one-piece, single blade squeegee has an ultra-hygienic design that minimizes areas for water and food soils to collect. This seamless, one-piece construction is especially effective in areas requiring the highest level of hygiene. The soft and flexible rubber blade is ideal for removing water or food debris from floors and food contact surfaces. Hygiene squeegees are a good foundation for any HACCP or food safety program.
Delivery: Dropshipping available.
Customers Who Bought This Also Bought