Tool Sets for Process Reactors, Chemglass
Tool Set for Process Reactor with 17 Piece
Catalog # 89028-858
Supplier: Chemglass
: CG-1963-B-30
Jump to:
Product Details & Documents
Precision crafted 17-piece tool set which includes a variety of tools to handle basic assembly, repair and maintenance on process reactor systems.
: Tool set includes beam torque wrench, 3/8" deep well socket, 7/16" deep well socket, 8 and 10" adjustable wrench set, allen/hex key caddy set, SAE and metric, 5/32" T-handle allen wrench, 3/16" T-handle allen wrench, 1/4" T-handle allen wrench, 10 in 1 screw driver, strap wrench, plastic tool box, tubing cutter for plastic and rubber tubing, Robogrip® pliers and plastic cushion jaws.
Spécifications
- Désignation:Tool Set for Process Reactor with 17 Piece
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- ³/₈" deep well socket, ¼" drive
Informations sur le produit
- Catalog #AvailabilityPrice
- ⁷/₁₆" deep well socket, ¼" drive
Informations sur le produit
- Catalog #AvailabilityPrice
- Beam torque wrench, ¼ drive, 11 overall lengthBeam torque wrench, ¼ drive, 11 overall length16002-878Log in pour la disponibilitéEach - $242.16
Informations sur le produit
- Catalog #AvailabilityPrice
- Adjustable wrench set, 8 and 10"
Informations sur le produit
- Catalog #AvailabilityPrice
- Allen/Hex key caddy set, SAE and metric
Informations sur le produit
- Catalog #AvailabilityPrice
- ³/₁₆" T-handle Allen wrench
Informations sur le produit
- Catalog #AvailabilityPrice
- ⁵/₃₂" T-handle Allen wrench
Informations sur le produit
- Catalog #AvailabilityPrice
- ¼" T-Handle Allen wrench
Informations sur le produit
- Catalog #AvailabilityPrice
- 10 in 1 screw driver
Informations sur le produit
- Catalog #AvailabilityPrice
- Strap wrench, 4 O.D. capacity
Informations sur le produit
- Catalog #AvailabilityPrice
- Plastic tool box, W×D×H: 19×10×9"
Informations sur le produit
- Catalog #AvailabilityPrice
- Robogrip® pliers, plastic cushion jaws, 2 piece setRobogrip® pliers, plastic cushion jaws, 2 piece set16002-898Log in pour la disponibilitéEach - $215.51
Informations sur le produit
- Catalog #AvailabilityPrice
- Tubing cutter for plastic and rubber tubing
Informations sur le produit
- Catalog #LengthWidthAvailabilityPrice
- PTFE Tape