Top Dog Large 6 Mil Gown, Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
Reusable and washable, the one-piece construction increases the gown's hygiene and durability.
- Resistance to chemical and acid sanitation products
- Non-porous surface will not host bacteria or mold
- Washer and dryer safe
- Will not crack or stiffen in low temperatures or with repeated cleaning
- Impervious to animal fats and degreasers
- Completely latex-free for sensitive users
- Easily repairable
Made of one-piece polyurethane, the 6 Mil Gown offers supreme protection for those in the food industry. There are no strings or hardware, which are a contamination risk. This gown is chemical and abrasive resistant, and machine washable and dryer-safe. All gowns featured in this collection are size Large garments. The full-length sleeves are cuffed for extra protection. Along with the hygienic nature of the garment itself and the five colors it comes in to complement Remco and Vikan's main product line colors, the 6 Mil Gown is ideal for protection in food processing environments.
Certifications: Made of FDA Compliant material.
Ordering information: Gowns can only be purchased by the case. Case quantity is 10.
Delivery: Dropshipping available.
Spécifications
- Catalog No:
- 76196-548
- 76196-546
- 76196-552
- 76196-550
- 76196-554
- Garment application:
- Food, Chemical protection
- Food, Chemical protection
- Food, Chemical protection
- Food, Chemical protection
- Food, Chemical protection
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Fabric:
- Ether-based polyurethane
- Ether-based polyurethane
- Ether-based polyurethane
- Ether-based polyurethane
- Ether-based polyurethane
- Thickness:
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil