Top Dog 6 Mil Sleeves, 21" Length, Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
Reusable and washable, the one-piece construction increases the sleeve's hygiene and durability.
- Resistance to chemical and acid sanitation products
- Non-porous surface will not host bacteria or mold
- Washer and dryer safe
- Will not crack or stiffen in low temperatures or with repeated cleaning
- Impervious to animal fats and degreasers
- Completely latex-free for sensitive users
- Easily repairable
These sleeves are made of 6 mil pure polyurethane. They provide coverage from above the elbow to the wrist, with elastic at both ends to keep them in place. Five colors make the sleeves coordinate with Top Dog's aprons and Remco and Vikan's standard product line colors. The sleeves add extra coverage while providing worker protection and flexibility, both in design and in function.
Certifications: Made of FDA Compliant material.
Ordering information: Sleeves can only be purchased by the case. Case quantity is 18.
Delivery: Dropshipping available.
Spécifications
- Catalog No:
- 76196-544
- 76196-538
- 76196-536
- 76196-542
- 76196-540
- Garment application:
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Fabric:
- Ether-based polyurethane
- Ether-based polyurethane
- Ether-based polyurethane
- Ether-based polyurethane
- Ether-based polyurethane
- Thickness:
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil