ALPS™ Heat Sealer, Thermo Scientific
Catalog # 89499-572
Supplier: Thermo Fisher Scientific
Specifications
- Description:Semi automated microplate heat sealer
- Dimensions:220×425 mm
- Operating humidity range:20 - 80% noncondensing
- Operating temperature:18...30 °C
- Cat. no.:89499-572
- Electrical:100 - 130 V
- Weight:7.2 kg
- Supplier No.:AB-1443A
Specifications
About this item
ALPS™ Microplate heat sealer is designed to provide sealing consistency in low to medium throughput labs.
- Offers superior sample protection of storage and reaction plates
- Applications includes compound storage, sample archiving and PCR
- Simple on-board touch pad controls adjust sealing temperature and time for reproducible results
- Audible alarm indicates sealing completion
- Lighweight: 7.2 kg
Apply consistent, secure, tight seals around individual wells, eliminating sample loss through evaporation and cross contamination between wells.
Counter lever handle design provides minimal strain when operating the instrument.
Customers Who Bought This Also Bought