Twirl'Blue PE Gloves, Sterile, Labplas
# de catalogue 75849-336
Fournisseur: Labplas
Spécifications
- Food contact approved:Yes
- Glove Color:Blue
- Description:Twirl'Blue gloves, gamma sterilized
- Disposable / reusable:Disposable
- Cat. no.:75849-336
- Packaging:50 Pairs/Box
- Supplier No.:SA7-GLOVES
Spécifications
A propos de cet article
These gamma sterilized blue color gloves are made of polyethylene and specifically designed for food industries to avoid contaminations.
- R-Nase, D-Nase and pyrogen free
- Accommodate solid, semi-solid and liquid compounds
- Food contact approved
The main purpose of the distinct color design is to avoid possible foreign matter contamination and for increased visibility.
Customers Who Bought This Also Bought