Dippers, PTFE, Dynalon
Supplier: Dynalon
Certificates
About this item
Totally inert and non-contaminating PTFE dipper sampler.
Extendable screw-in shafts have a steel core for rigidity and are fully encapsulated in PTFE.
Delivery: The basic unit consists of a beaker with a single 24" (600 mm) shaft. Replacement shaft is available separately.
Customers Who Bought This Also Bought