Giant Classroom Thermometer
# de catalogue 470237-802
Fournisseur: Learning Resources
Spécifications
- Description:Giant Classroom Thermometer
- Cat. no.:470237-802
- Range:– 40...+50 °C (– 40...+120 °F)
Spécifications
A propos de cet article
A great choice for students of all ages.
- Wall-mounted thermometer
- Recessed tube prevents breakage
- Oversized design ideal for easy reading
Largest working thermometer available for class display! The 30" wall-mounted thermometer has a 23" tube recessed into the sturdy wooden base.
Customers Who Bought This Also Bought