Liquid Sampler, HDPE, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
This sampling device is perfect for corrosive environments and for hard to reach places.
- Excellent for industrial and environmental applications
Plastic Liquid Sampler is manufactured from chemically resistant high density polyethylene.
The included bottles screw into the end of the handle and is filled via four 5mm diameter ports (2 allowing liquid to enter and 2 allowing air to escape). All rods are 1.25" O.D.
Packaging: Two-piece handle for UPS shipment.
Customers Who Bought This Also Bought