Oakton® pH Testr™ 50 Waterproof Pocket pH Tester, Premium 50 Series, Cole-Parmer
Catalog # 76208-088
Supplier: Argos Technologies
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Calibration:Up to 3 points
- Temperature range:0 – 60 °C (32 – 140 °F)
- Description:Waterproof Pocket pH Tester
- Temperature accuracy:±0.5 °C (±0.9 °F)
- Temperature resolution:0.1 °C (0.1 °F)
- Battery life:250 hrs.
- Automatic buffer recognition:USA 4.01, 7.00, 10.01 and NIST 4.01, 6.86, 9.18
- Temperature compensation:ATC
- pH accuracy:±0.01 pH
- pH range:–1 to 15
- pH resolution:0.01 pH
- Battery type:Four AAA 1.5 V
- Cat. no.:76208-088
Specifications
About this item
Versatile, compact design works in the field or laboratory.
- Easy-to-navigate interface for quick setup and calibration
- Replaceable, double-junction electrode sensors save time and money
- Automatic temperature compensation ensures accurate reading in varying environments
- Waterproof
- LCD display
Redesigned leakproof cap functions as a sample cup, hydrating cup and stabilizing base to stand the meter upright.
Ordering information: Includes protective plastic storage case, lanyard, and 4 AAA batteries.
Frequently Bought Together