ContactUsLinkComponent
Accessories for Sash-Glider Kit, Labconco
Accessories for Sash-Glider Kit, Labconco
# de catalogue 10127-518
Fournisseur:  Labconco
Product available on GSA Advantage®
Sash-glider scrubbing strip weight: 2. 0 lbs, weight metric: 1. 0 kg
Accessories for Sash-Glider Kit, Labconco
# de catalogue 10127-518
Fournisseur:  Labconco
Product available on GSA Advantage®
Spécifications
  • Description:
    Refill Pack of 25 Sash-Glider Scrubbing Strips
  • Cat. no.:
    10127-518
  • Supplier No.:
    3408600