About this item
Sklar Edge™ TC Lexer scissors can be used in multiple procedures and settings.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
Sklar Edge™ TC Lexer scissors have a standard beveled blade. Depending on physician preference, both the straight and the curved version may be used interchangeably, but the straight version is typically used for cutting surface tissue or sutures. Sklar Edge™ Tungsten Carbide Scissors provide an exceptionally durable and hard cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than standard scissors. Sklar permanently bonds tungsten carbide inserts to the blades of the scissors which maximizes control and precision. They can be identified by two gold coated finger rings.