Spécifications
- Length:5"
- Material:Stainless Steel
- Cat. no.:76192-330
Spécifications
A propos de cet article
Econo™ Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
This handle has horizontal serrations for thumb and forefinger gripping. The #3 handle has centimeter and millimeter markings on the handle for measuring tissue. Econo™ instruments are Sklar's floor grade, disposable instruments and are sold in bulk quantities.