Scalpel-less Dissection Kit
Catalog # 470234-476
Supplier: DR INSTRUMENTS INC. SE
Specifications
- Description:Dissection Kit
- Cat. no.:470234-476
- Includes:5 Instruments
- No. of Parts:7
- No. of Lab Groups:1
Specifications
About this item
Great tools for a safer dissection.
- High-quality instruments
- Stainless steel resists corrosion
Practice dissection with a lower chance of dangerous cuts! Ideal for beginners in dissection, where exposed scalpel blades may be a hazard. This scalpel-free dissection kit has the tools needed to perform most dissections in a classroom. Suitable for most gross dissection and introductory organisms like squid, worms, grasshoppers, and clams. Each of the instruments is the same as you will find in our other high-quality kits and will provide years of service.
Packaging: Please see datasheet for contents.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Customers Who Bought This Also Bought