Compact Refrigerated Category B Shipping Systems, Therapak®
Fournisseur: Therapak, LLC
Certificats
A propos de cet article
Compact refrigerated shippers are an ideal solution to meet the demands of medical and clinical specimen shipping.
- Easy to use
- Category B certified
- Ship up to six 10 mL or smaller Blood Tubes
- 95 kPa pressure rated secondary packaging
- Complete shipping system
These certified Category B Shippers transport up to six 10 mL blood tubes. Precondition the included refrigerant packs for 24 hours at -20 ˚C and place in the cooler with the specimens. These shippers have been certified to transport Category B Specimens.
Certifications: Meets IATA Packing Instructions 650 requirements to transport Category B Specimens.
Customers Who Bought This Also Bought