Metal Detectable Earplugs, Moldex®
Fournisseur: Moldex/Metric
Certificats
A propos de cet article
Moldex® Metal detectable earplugs are perfect for food processing and other applications that require detectability. Cords are securely attached to the plugs. All Moldex® hearing protection is independently tested for protection that can be counted on.
- Moldex® Metal detectable corded earplugs are available in reusable Rockets® or soft foam SparkPlugs®
- Both models feature a securely attached metal detectable cord and a bright blue color that is ideal for any hearing protection program, especially in the food processing industry
- SparkPlugs® are rated NRR 33, the highest independently tested NRR available for use in the most noisiest environments
- Rockets® feature an independently tested NRR 27 and can be washed and reused
- SparkPlugs® are made of extra-soft foam and have a tapered design to fit snugly and comfortably in the ear
- SparkPlugs® have an enclosed metal ball bearing which eliminates any exposed metal
- Rockets® have an enclosed metal spring which eliminates any exposed metal
- The ball bearing or spring are in seperate holes ensuring the plugs are still detectable even if the cord is removed
- Independently tested to ANSI S3.19-1974
- 100% PVC-free, like all Moldex® products and packaging, offering a greener alternative
- Made in U.S.A.
Spécifications
- Catalog No:
- 89011-750
- 56219-250
- Environmentally Preferable:
- Sustainable Packaging
- Sustainable Packaging