ReadyStrain™ Sterile Cell Straining Kits, MTC Bio
Fournisseur: Avantor
Certificats
A propos de cet article
ReadyStrain™ is the first product of its kind, providing all the necessary parts for filtration of cell suspensions and combining them into a single, pre-assembled, ready to use, sterile packet.
- Pre-assembled & supplied ready to use
- Opens directly under the hood to reduce contamination risks
- Garaunteed sterile via gamma sterilization
- Advanced design provides faster flow-through
These kits eliminate the contamination risks associated with opening multiple packages, and assembling separate pieces (i.e. tubes, strainers, and caps). The individual sterile packets mean researchers are no longer forced to expose entire bags of 50 mL tubes and corresponding caps in order to perform one filtration. Instead they can open a single ReadyStrain™ packet directly under the hood. Processing and storage after the filtration is complete.
Delivery: Pre-assembled sterile cell straining kits include a premium cell strainer, a 50 mL tube, and separate cap.
Packaging: Packaged in easy-open "tear-strip" bags.
Customers Who Bought This Also Bought