SP Bel-Art Protective Tapes, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Tapes made of PP provides great versatility for heavy-duty lab use.
Tapes protect labels from the freezer, autoclave, acids, alkalis and organic solvents.
Customers Who Bought This Also Bought