SP Bel-Art Labels, Bel-Art Products, a part of SP
# de catalogue 36440-640
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Color:White
- Description:Labels
- Cat. no.:36440-640
- Length:3"
- Width:1"
- Supplier No.:F13455-0010
Spécifications
A propos de cet article
Labels accept any kind of marker. Perfect for use under the clear tape to label labware, bottles or samples.
Customers Who Bought This Also Bought