CoreSafe® Natural Rubber Chemical-Resistant Gloves, Unlined, SW Safety Solutions
Fournisseur: SW Safety Solutions
Certificats
A propos de cet article
A natural color latex hand protection solution in a 17 mil palm thickness with a 12" length.
- Fish scale grip pattern
- Flock-lined for donning and comfort
- Straight cut cuff
- Imprinted cuff
- Double chlorinated inner and outer surfaces
The fish scale grip pattern on this hand-specific, unsupported chemical resistant glove provides advanced grip and protection in a variety of applications.
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; SHA Dermatologically Approved; EAC TP TC 019/2011; CE 0493.
Ordering information: Glove sizes are as follows; S (7), M (8), L (9), XL (10).
Packaging: Gloves are packaged 12 pairs per bag (144 pairs per case).
Customers Who Bought This Also Bought