Progressive Spatula Set
# de catalogue 470089-212
Fournisseur: PROGRESSIVE INTERNATIONAL CORP
Spécifications
- Color:White
- Description:Progressive rubber spatula set, set of 3
- Cat. no.:470089-212
- Supplier No.:GT-3323
Spécifications
A propos de cet article
The progressive set of three spatulas is ideal for mixing and scraping batter or egg whites, or spreading frosting.
- Dishwasher safe
- Features plastic handle
Delivery: Set includes: 2.25" spoon spatula, 2" spatula, and 1.25" spatula.
Customers Who Bought This Also Bought