Frosted Slides, Azer Scientific
# de catalogue 76076-738
Fournisseur: Azer Scientific Inc.
Spécifications
- Description:Single-frosted slides
- L×W:75×25 mm
- Thickness:1.2 mm
- Cat. no.:76076-738
Spécifications
A propos de cet article
Smooth slides with burr-free, wet-ground edges are frosted at a single end in order to prevent jamming and sticking.
Packaging: Slides are packaged 72 per box, 20 boxes per case.
Customers Who Bought This Also Bought