Fume Hood Support Kits
Supplier: OHAUS CORP.
Certificates
About this item
Provides versatile secure support for holding laboratory glassware.
- Stainless steel construction
- Three sizes available to fit most fume hood needs
Choose from three kits specifically designed to fit within laboratory fume hoods. All components are constructed of stainless steel with the exception of the Frame Connectors which are made of aluminum with a silicon finish. 4' (1.22 m) kit constructs frame that is 36 x 38" (914 x 965 mm); 5' (1.52 m) kit constructs frame that is 36 x 50" (914 x 1270 mm); 6' (1.83 m) kit constructs frame that is 36 x 62" (914 x 1575 mm). All bases are 12" (305 mm) wide.
Customers Who Bought This Also Bought