Westergren Erythrocyte Sedimentation Rate Tubes, KIMBLE CHASE®, DWK Life Sciences
Catalogue # 60826-605
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Ø:2.55 mm
- Description:Westergren Erythrocyte Sedimentation Rate Tubes
- Length:240 mm
- Cat. no.:60826-605
- Supplier No.:67G830SO
Spécifications
À propos de cet article
ESR tubes measure the rate at which red blood cells settle out of plasma.
- Calibrated in yellow
- Cotton plugged
Westergren sedimentation rate tubes may be used for standard procedures, by drawing blood in a liquid sodium citrate vacuum tube, or the modified procedure, using a saline-diluted EDTA vacuum tube.
Precision-drawn glass and accurate calibrations ensure that the results are accurate with standard normal values.
Certifications: Manufactured from 90 expansion soda lime glass conforming to USP Type III requirements.