Heat Exchangers for Bioreactors, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Heat exchangers are available in a choice of two different styles.
The cold finger style is for use with 3 and 5 litre reactors and fits into the headplate via an M18 threaded port. The second style is a loop type exchanger and is available for 5 - 15 litre reactors. It fits into two, M10 ports on the headplate.
Customers Who Bought This Also Bought