Five-Neck Reaction Vessel Lids, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
These five neck reaction vessel lids have a flat ground surface allowing it to be greased or used with an o-ring.
Three side necks are vertical and one neck has a 10° angle. Angled side neck is designed for use with angled distillation adapters.
Customers Who Bought This Also Bought