KIMAX® Funnels, Long Stem, Thistle Top, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Funnels are manufactured from low extractable borosilicate glass.
- Funnels are cylindrical in shape
- 12" long stem, thistle top funnels
Certifications: Designed from USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought