Nonsterile New Sponge Dressings, DUKAL™ Corporation
Fournisseur: Dukal
Certificats
A propos de cet article
Nonsterile new sponges are a rayon/poly blend construction that are virtually non-linting.
- Rayon/poly blend construction
- The effective combination of weight and open aperture allows each sponge to wick quicker and offer superior absorbency
- Virtually non-linting
- Not made with natural rubber latex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.